CH6371380493 Chem Impex International, Inc.

(R)-3-(Boc-amino)-2-oxo-1-pyrrolidine-acetic acid

  • MDL number MFCD02684393
  • Cas Number : 78444-90-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 99
Storage temp. Store at 0-8
Physical Form White solid
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-r-i-3-boc-amino-2-oxo-1-pyrrolidine-acetic-acid/5540?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-r-i-3-boc-amino-2-oxo-1-pyrrolidine-acetic-acid/5540?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart