CH6371380494 Chem Impex International, Inc.

Boc-(S)-3-amino-2-oxo-1-pyrrolidine-acetic acid

  • MDL number MFCD02684392
  • Cas Number : 79839-26-2
  • Molecular Weight : 258.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Store at 0-8
Physical Form White to yellow solid
Shipping Temperature RT
Country of Origin US
Product Link http://www.chemimpex.com/product/productinfo/boc-i-s-i-3-amino-2-oxo-1-pyrrolidine-acetic-acid/5543?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/boc-i-s-i-3-amino-2-oxo-1-pyrrolidine-acetic-acid/5543?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart