CH6371380495 Chem Impex International, Inc.

1-tert-Butoxy-3-propylamine

  • MDL number MFCD02684390
  • Cas Number : 100364-10-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 3-Aminopropyl tert-butyl ether
Purity 99
Storage temp. Store at 0-8
Physical Form Clear liquid
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/1-i-tert-i-butoxy-3-propylamine/5545?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/1-i-tert-i-butoxy-3-propylamine/5545?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart