CH6371380500 Chem Impex International, Inc.

Acetyl-D-homophenylalanine

  • MDL number MFCD00190716
  • Cas Number : 63393-59-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 174-179
Synonym Ac-D-HomoPhe-OH
Purity 99
Storage temp. Store at 0-8
Physical Form White crystals
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/acetyl-d-homophenylalanine/5552?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/acetyl-d-homophenylalanine/5552?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart