CH6371380501 Chem Impex International, Inc.

Acetyl-DL-glutamic acid

  • MDL number MFCD00063195
  • Cas Number : 5817-08-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Ac-DL-Glu-OH
Purity 98
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/acetyl-dl-glutamic-acid/5553?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/acetyl-dl-glutamic-acid/5553?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart