CH6371380502 Chem Impex International, Inc.

Acetyl-DL-norleucine

  • MDL number MFCD00037272
  • Cas Number : 7682-16-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 99
Storage temp. Store at 0-8°C
Physical Form White powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/acetyl-dl-norleucine/5555?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/acetyl-dl-norleucine/5555?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart