CH6371380504 Chem Impex International, Inc.

Acetyl-D-norleucine

  • MDL number MFCD00063171
  • Cas Number : 54896-21-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 101-108
Synonym Ac-D-Nle-OH
Purity 99
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/acetyl-d-norleucine/5559?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/acetyl-d-norleucine/5559?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart