CH6371380505 Chem Impex International, Inc.

Acetyl-D-norvaline

  • MDL number MFCD00151962
  • Cas Number : 57357-56-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 98-104
Synonym Ac-D-Nva-OH
Purity 99
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/acetyl-d-norvaline/5560?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/acetyl-d-norvaline/5560?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart