CH6371380506 Chem Impex International, Inc.

Acetyl-3-cyano-D-phenylalanine

  • MDL number MFCD01862349
  • Cas Number : 599178-70-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Ac-D-Phe(3-CN)-OH
Storage temp. Store at 0-8
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/acetyl-3-cyano-d-phenylalanine/5561?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/acetyl-3-cyano-d-phenylalanine/5561?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart