CH6371380507 Chem Impex International, Inc.

Acetyl-4-chloro-D-phenylalanine

  • MDL number MFCD00672506
  • Cas Number : 135270-40-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 119-125
Synonym Ac-D-Phe(4-Cl)-OH
Purity 99
Storage temp. Store at 0-8
Physical Form White crystalline powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/acetyl-4-chloro-d-phenylalanine/5562?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/acetyl-4-chloro-d-phenylalanine/5562?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart