CH6371380515 Chem Impex International, Inc.

Acetyl-L-norvaline

  • MDL number MFCD00151963
  • Cas Number : 15891-50-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 96-106
Synonym Ac-L-Nva-OH
Purity 98
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/acetyl-l-norvaline/5571?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/acetyl-l-norvaline/5571?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart