CH6371380517 Chem Impex International, Inc.

N,O-Bis-acetyl-L-tyrosine

  • MDL number MFCD00038242
  • Cas Number : 17355-23-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 162-168 �C
Synonym Ac-L-Tyr(Ac)-OH
Purity 98
Storage temp. Store at 0-8
Physical Form White crystalline powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-i-o-i-bis-i-i-acetyl-l-tyrosine/5573?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-i-o-i-bis-i-i-acetyl-l-tyrosine/5573?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart