CH6371380518 Chem Impex International, Inc.

Acetyl-O-methyl-L-tyrosine

  • MDL number MFCD00026170
  • Cas Number : 28047-05-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Ac-L-Tyr(Me)-OH
Storage temp. Store at 0-8
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/acetyl-i-o-i-methyl-l-tyrosine/5574?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/acetyl-i-o-i-methyl-l-tyrosine/5574?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart