CH6371380524 Chem Impex International, Inc.

Boc-Asn-Gly-OEt

  • MDL number MFCD00270491
  • Cas Number : 211056-85-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 144-149
Synonym Nalpha-Boc-asparagine glycine ethyl ester
Purity 98
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/boc-asn-gly-oet/5582?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/boc-asn-gly-oet/5582?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart