CH6371380525 Chem Impex International, Inc.

Boc-L-aspartic acid b-methyl ester dicyclohexylammonium salt

  • MDL number MFCD00270494

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 155-162
Synonym Boc-L-Asp(OMe)-OH�A
Purity 98
Storage temp. Store at 0-8
Physical Form White solid
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/boc-l-aspartic-acid-b-methyl-ester-dicyclohexylammonium-salt/5583?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/boc-l-aspartic-acid-b-methyl-ester-dicyclohexylammonium-salt/5583?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart