CH6371380532 Chem Impex International, Inc.

Na-Boc-Ng-allyloxycarbonyl-D-2,4-diaminobutyric acid dicyclohexylammonium salt

  • MDL number MFCD02684370
  • Cas Number : 350820-59-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 118-127
Synonym Boc-D-Dab(Aloc)-OH DCHA
Purity 99
Storage temp. Store at 0-8
Physical Form White crystalline powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-sup-alpha-sup-i-i-boc-i-n-i-sup-gamma-sup-allyloxycarbonyl-d-2-4-diaminobutyric-acid-dicyclohexylammonium-salt/5590?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-sup-alpha-sup-i-i-boc-i-n-i-sup-gamma-sup-allyloxycarbonyl-d-2-4-diaminobutyric-acid-dicyclohexylammonium-salt/5590?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart