CH6371380533 Chem Impex International, Inc.

Boc-D-glutamic acid gamma-allyl ester

  • MDL number MFCD01862283
  • Cas Number : 259221-91-5
  • Molecular Weight : 287.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 43-49 °C
Purity 99
Storage temp. Store at 0-8°C
Physical Form White solid
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/boc-d-glutamic-acid-allyl-ester/5591?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/boc-d-glutamic-acid-allyl-ester/5591?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart