CH6371380536 Chem Impex International, Inc.

Boc-D-phenylglycine N-hydroxysuccinimide ester

  • MDL number MFCD00270515
  • Cas Number : 39249-27-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 166-173
Synonym Boc-D-Phg-OSu
Purity 99
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/boc-d-phenylglycine-i-n-i-hydroxysuccinimide-ester/5594?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/boc-d-phenylglycine-i-n-i-hydroxysuccinimide-ester/5594?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart