CH6371380537 Chem Impex International, Inc.

Boc-D-serine benzyl ester

  • MDL number MFCD00080265
  • Cas Number : 141527-78-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 66-70 �C
Synonym Boc-D-Ser-OBzl
Purity 99
Storage temp. Store at 0-8
Physical Form White to off-white powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/boc-d-serine-benzyl-ester/5595?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/boc-d-serine-benzyl-ester/5595?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart