CH6371380539 Chem Impex International, Inc.

Boc-O-allyl-D-tyrosine

  • MDL number MFCD01862284
  • Cas Number : 350820-56-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 68-81
Synonym Boc-D-Tyr(All)-OH
Purity 99
Storage temp. Store at 0-8
Physical Form White crystalline powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/boc-i-o-i-allyl-d-tyrosine/5597?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/boc-i-o-i-allyl-d-tyrosine/5597?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart