CH6371380541 Chem Impex International, Inc.

Boc-O-methyl-L-homoserine dicyclohexylammonium salt

  • MDL number MFCD01861359
  • Cas Number : 349545-91-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 136-143
Synonym Boc-L-HomoSer(Me)-OH DCHA
Purity 99
Storage temp. Store at 0-8
Physical Form White to off-white crystals
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/boc-i-o-i-methyl-l-homoserine-dicyclohexylammonium-salt/5601?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/boc-i-o-i-methyl-l-homoserine-dicyclohexylammonium-salt/5601?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart