CH6371380543 Chem Impex International, Inc.

Boc-L-6-hydroxynorleucine

  • MDL number MFCD01862292
  • Cas Number : 77611-37-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 108-119
Synonym Boc-L-Nle(6-OH)
Purity 99
Storage temp. Store at 0-8
Physical Form White crystalline powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/boc-l-6-hydroxynorleucine/5603?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/boc-l-6-hydroxynorleucine/5603?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart