CH6371380547 Chem Impex International, Inc.

Nalpha-Boc-Nepsilon-isopropyl-L-lysine

  • MDL number MFCD00270511
  • Cas Number : 66880-55-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 99
Storage temp. Store at 0-8°C
Physical Form White powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-sup-a-sup-boc-i-n-i-sup-e-sup-isopropyl-l-lysine/5607?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-sup-a-sup-boc-i-n-i-sup-e-sup-isopropyl-l-lysine/5607?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart