CH6371380548 Chem Impex International, Inc.

Boc-L-norvaline N-hydroxysuccinimide ester

  • MDL number MFCD01862291
  • Cas Number : 108233-37-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 91-96 °C
Purity 98
Storage temp. Store at 0-8°C
Physical Form White to off-white powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/boc-l-norvaline-i-n-i-hydroxysuccinimide-ester/5608?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/boc-l-norvaline-i-n-i-hydroxysuccinimide-ester/5608?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart