CH6371380554 Chem Impex International, Inc.

Boc-Nin-formyl-L-1,2,3,4-tetrahydronorharmane-3-carboxylic acid

  • MDL number MFCD01862297
  • Cas Number : 327603-46-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 113-125
Synonym Boc-L-Tpi(For)-OH
Purity 99
Storage temp. Store at 0-8
Physical Form White crystalline powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/boc-i-n-i-sup-in-sup-formyl-l-1-2-3-4-tetrahydronorharmane-3-carboxylic-acid/5614?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/boc-i-n-i-sup-in-sup-formyl-l-1-2-3-4-tetrahydronorharmane-3-carboxylic-acid/5614?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart