CH6371380560 Chem Impex International, Inc.

Benzoyl-DL-leucine

  • MDL number MFCD00038295
  • Cas Number : 17966-67-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 140-144 °C
Purity 99
Storage temp. Store at 0-8°C
Physical Form White to off-white solid
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/benzoyl-dl-leucine/5622?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/benzoyl-dl-leucine/5622?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart