CH6371380561 Chem Impex International, Inc.

Na-Benzyl-L-alanine methyl ester hydrochloride

  • MDL number MFCD02684375
  • Cas Number : 19460-85-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 181-187
Synonym Bzl-L-Ala-OMe HCl
Purity 99
Storage temp. Store at 0-8
Physical Form White crystalline powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-sup-alpha-sup-benzyl-l-alanine-methyl-ester-hydrochloride/5623?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-sup-alpha-sup-benzyl-l-alanine-methyl-ester-hydrochloride/5623?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart