CH6371380563 Chem Impex International, Inc.

Na-Benzyl-D-alanine methyl ester hydrochloride

  • MDL number MFCD02684374
  • Cas Number : 95071-12-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 178-186
Synonym Bzl-D-Ala-OMe HCl
Purity 99
Storage temp. Store at 0-8
Physical Form White crystalline powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-sup-alpha-sup-benzyl-d-alanine-methyl-ester-hydrochloride/5625?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-sup-alpha-sup-benzyl-d-alanine-methyl-ester-hydrochloride/5625?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart