CH6371380564 Chem Impex International, Inc.

N-Benzyl-L-glutamine methyl ester hydrochloride

  • MDL number MFCD02684377
  • Cas Number : 402929-52-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 125-132 °C
Purity 99
Storage temp. Store at 0-8 °C
Physical Form White crystalline powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-benzyl-l-glutamine-methyl-ester-hydrochloride/5626?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-benzyl-l-glutamine-methyl-ester-hydrochloride/5626?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart