CH6371380566 Chem Impex International, Inc.

Benzyl-L-isoleucine methyl ester hydrochloride

  • MDL number MFCD02684380

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 176-182
Synonym Benzyl-L-isoleucine methyl ester hydrochloride
Purity 99
Storage temp. Store at 0-8
Physical Form White crystalline powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/benzyl-l-isoleucine-methyl-ester-hydrochloride/5628?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/benzyl-l-isoleucine-methyl-ester-hydrochloride/5628?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart