CH6371380567 Chem Impex International, Inc.

Benzyl-L-methionine methyl ester hydrochloride

  • MDL number MFCD02684382
  • Cas Number : 402929-60-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 92-99 °C
Purity 99
Storage temp. Store at 0-8 °C
Physical Form White crystalline powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/benzyl-l-methionine-methyl-ester-hydrochloride/5629?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/benzyl-l-methionine-methyl-ester-hydrochloride/5629?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart