CH6371380568 Chem Impex International, Inc.

Benzyl-L-phenylalanine methyl ester hydrochloride

  • MDL number MFCD02684383
  • Cas Number : 7703-09-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 142-155 °C
Purity 99
Storage temp. Store at 0-8 °C
Physical Form White crystalline powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/benzyl-l-phenylalanine-methyl-ester-hydrochloride/5630?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/benzyl-l-phenylalanine-methyl-ester-hydrochloride/5630?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart