CH6371380571 Chem Impex International, Inc.

Benzoyl-L-norleucine

  • MDL number MFCD00190768
  • Cas Number : 54430-46-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 91-99 �C
Synonym Bz-L-Nle-OH
Purity 99
Storage temp. Store at 0-8
Physical Form White crystalline powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/benzoyl-l-norleucine/5633?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/benzoyl-l-norleucine/5633?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart