CH6371380572 Chem Impex International, Inc.

Benzoyl-L-norvaline

  • MDL number MFCD00190769
  • Cas Number : 121470-62-0
  • Molecular Weight : 221.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 83 - 89 °C
Purity 98
Storage temp. Store at 0-8°C
Physical Form Off-white powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/benzoyl-l-norvaline/5634?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/benzoyl-l-norvaline/5634?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart