CH6371380573 Chem Impex International, Inc.

Chloroacetyl-DL-2-amino-N-butyric acid

  • MDL number MFCD00055800
  • Cas Number : 67183-18-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 130-136 °C
Purity 99
Storage temp. Store at 0-8°C
Physical Form White powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/chloroacetyl-dl-2-amino-i-n-i-butyric-acid/5635?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/chloroacetyl-dl-2-amino-i-n-i-butyric-acid/5635?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart