CH6371380574 Chem Impex International, Inc.

Chloroacetyl-DL-norvaline

  • MDL number MFCD00020439
  • Cas Number : 6940-47-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 96-101 �C
Synonym ClAc-DL-Nva-OH
Purity 99
Storage temp. Store at 0-8
Physical Form Off-white powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/chloroacetyl-dl-norvaline/5636?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/chloroacetyl-dl-norvaline/5636?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart