CH6371380575 Chem Impex International, Inc.

Chloroacetyl-L-norleucine

  • MDL number MFCD00190854
  • Cas Number : 56787-36-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 67-72 °C
Purity 99
Storage temp. Store at 0-8°C
Physical Form White powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/chloroacetyl-l-norleucine/5637?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/chloroacetyl-l-norleucine/5637?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart