CH6371380578 Chem Impex International, Inc.

Fmoc-cis-4-aminocyclohexane carboxylic acid

  • MDL number MFCD01862344
  • Cas Number : 147900-45-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 131-138
Synonym Fmoc-1,4-cis-Achc-OH
Purity 99
Storage temp. Store at 0-8
Physical Form White crystalline powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/fmoc-i-cis-i-4-aminocyclohexane-carboxylic-acid/5641?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/fmoc-i-cis-i-4-aminocyclohexane-carboxylic-acid/5641?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart