CH6371380580 Chem Impex International, Inc.

Fmoc-S-acetamidomethyl-L-cysteine N-hydroxysuccinimide ester

  • MDL number MFCD01076143
  • Cas Number : 182410-75-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 145-153
Synonym Fmoc-L-Cys(Acm)-OSu
Purity 98
Storage temp. Store at 0-8
Physical Form Off-white powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/fmoc-i-s-i-acetamidomethyl-l-cysteine-i-n-i-hydroxysuccinimide-ester/5643?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/fmoc-i-s-i-acetamidomethyl-l-cysteine-i-n-i-hydroxysuccinimide-ester/5643?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart