CH6371380581 Chem Impex International, Inc.

Fmoc-S-xanthyl-L-cysteine

  • MDL number MFCD01862337
  • Cas Number : 186829-25-4
  • Molecular Weight : 523.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 190-194 °C
Purity 98
Storage temp. Store at 0-8°C
Physical Form White to off-white powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/fmoc-i-s-i-xanthyl-l-cysteine/5644?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/fmoc-i-s-i-xanthyl-l-cysteine/5644?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart