CH6371380582 Chem Impex International, Inc.

Na-Fmoc-Ng-xanthyl-D-asparagine

  • MDL number MFCD01076135
  • Cas Number : 353236-19-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 211-217
Synonym Fmoc-Ngamma-xanthyl-D-asparagine
Purity 99
Storage temp. Store at 0-8
Physical Form White crystalline powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-sup-alpha-sup-i-i-fmoc-i-n-i-sup-gamma-sup-xanthyl-d-asparagine/5646?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-sup-alpha-sup-i-i-fmoc-i-n-i-sup-gamma-sup-xanthyl-d-asparagine/5646?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart