CH6371380583 Chem Impex International, Inc.

Nalpha-Fmoc-Ndelta-xanthyl-L-glutamine

  • MDL number MFCD01076141
  • Cas Number : 185031-81-6
  • Molecular Weight : 548.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 191-197 °C
Purity 99
Storage temp. Store at 0-8 °C
Physical Form White powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-sup-alpha-sup-i-i-fmoc-i-n-i-sup-delta-sup-xanthyl-l-glutamine/5647?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-sup-alpha-sup-i-i-fmoc-i-n-i-sup-delta-sup-xanthyl-l-glutamine/5647?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart