CH6371380585 Chem Impex International, Inc.

Fmoc-O-methyl-L-homoserine

  • MDL number MFCD00270543
  • Cas Number : 173212-86-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 138-143
Synonym Fmoc-L-HomoSer(Me)-OH
Purity 99
Storage temp. Store at 0-8
Physical Form White to off-white powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/fmoc-i-o-i-methyl-l-homoserine/5649?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/fmoc-i-o-i-methyl-l-homoserine/5649?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart