CH6371380586 Chem Impex International, Inc.

Fmoc-L-isoleucine N-hydroxysuccinimide ester

  • MDL number MFCD00190886
  • Cas Number : 121697-35-6
  • Molecular Weight : 450.49

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 73-81
Purity 98
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature RT
Country of Origin US
Product Link http://www.chemimpex.com/product/productinfo/fmoc-l-isoleucine-i-n-i-hydroxysuccinimide-ester/5650?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/fmoc-l-isoleucine-i-n-i-hydroxysuccinimide-ester/5650?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart