CH6371380590 Chem Impex International, Inc.

L-Asparagine 4-nitrobenzyl ester 4-toluenesulfonate salt

  • MDL number MFCD03788156
  • Cas Number : 3561-57-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 166-170
Synonym L-Asn-ONb TosOH
Purity 98
Storage temp. Store at 0-8
Physical Form Off-white powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/l-asparagine-4-nitrobenzyl-ester-4-toluenesulfonate-salt/5655?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/l-asparagine-4-nitrobenzyl-ester-4-toluenesulfonate-salt/5655?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart