CH6371380591 Chem Impex International, Inc.

L-Aspartic acid b-benzyl ester a-tert-butyl ester hydrochloride

  • MDL number MFCD00190761
  • Cas Number : 94347-11-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 109-119 �C
Synonym L-Asp(OBzl)-OtBu HCl
Purity 99
Storage temp. Store at 0-8
Physical Form White to off-white solid
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/l-aspartic-acid-beta-benzyl-ester-alpha-i-tert-i-butyl-ester-hydrochloride/5656?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/l-aspartic-acid-beta-benzyl-ester-alpha-i-tert-i-butyl-ester-hydrochloride/5656?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart