CH6371380594 Chem Impex International, Inc.

S-4-Methoxyltrityl-L-cysteine

  • MDL number MFCD05663448
  • Cas Number : 177582-20-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 160-165
Synonym L-Cys(Mmt)-OH
Purity 97
Storage temp. Store at 0-8
Physical Form Off-white powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-s-i-4-methoxyltrityl-l-cysteine/5659?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-s-i-4-methoxyltrityl-l-cysteine/5659?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart