CH6371380596 Chem Impex International, Inc.

D-Glutamic acid gamma-benzyl ester alpha-amide hydrochloride

  • MDL number MFCD05663749
  • Cas Number : 18800-75-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 180-187 °C
Purity 99
Storage temp. Store at 0-8 °C
Physical Form White crystalline powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/d-glutamic-acid-gamma-benzyl-ester-alpha-amide-hydrochloride/5661?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/d-glutamic-acid-gamma-benzyl-ester-alpha-amide-hydrochloride/5661?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart