CH6371380599 Chem Impex International, Inc.

DL-Serine benzyl ester 4-toluenesulfonate salt

  • MDL number MFCD00191018
  • Cas Number : 222739-29-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 98-105 �C
Synonym DL-Ser-OBzl TosOH
Purity 99
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/dl-serine-benzyl-ester-4-toluenesulfonate-salt/5666?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/dl-serine-benzyl-ester-4-toluenesulfonate-salt/5666?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart