CH6371380608 Chem Impex International, Inc.

trans-L-4-Hydroxyproline 4-nitroanilide hydrochloride

  • MDL number MFCD00190930

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 247 - 258 °C
Purity 99
Storage temp. Store at 0 - 8 °C
Physical Form Off-white or white crystalline powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-trans-i-l-4-hydroxyproline-4-nitroanilide-hydrochloride/5677?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-trans-i-l-4-hydroxyproline-4-nitroanilide-hydrochloride/5677?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart